Learn More
Cayman Chemical 2-Iminobiotin, Purity: ≥98%, CAS: 13395-35-2, 50 mg
SDP
Catalog No. 502983841
Encompass_Preferred
A cyclic guanidino analog of biotin that acts as a reversible inhibitor of iNOS and nNOS (Kis = 21.8 and 37.5 µM for mouse iNOS and rat nNOS, respectively)
Catalog No. 50-298-3841 Supplier Cayman Chemical Supplier No. 2081250 MG
A cyclic guanidino analog of biotin that acts as a reversible inhibitor of iNOS and nNOS (Kis = 21.8 and 37.5 µM for mouse iNOS and rat nNOS, respectively)