Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-iminobiotin hydrobromide | 76985-52-9 | 98.0% | 324.24 g·mol⁻1 | C10H18BrN3O2S | 1 ML
SDP

Catalog No. 5000228416
Encompass_Preferred

2-Iminobiotin hydrobromide is a biotin analog and a reversible inhibitor of nitric oxide synthases used in research to probe NO signaling and neuroprotection. It is supplied as a 10 mM solution in DMSO (1 mL) and has reported Ki values of 21.8 μM for murine iNOS and 37.5 μM for rat n-cNOS. For research use only; not for human or therapeutic applications.

  • Biotin analog that reversibly inhibits nitric oxide synthases (NOS).
  • Reported Ki: 21.8 μM (murine iNOS) and 37.5 μM (rat n-cNOS).
  • Supplied as 10 mM solution in DMSO, 1 mL vial.
  • Molecular weight 324.24 g·mol⁻1; formula C10H18BrN3O2S.
  • Purity: 98.0%.
  • Suitable for research on NO signaling, immunology, and neuroprotection.

Catalog No. 50-002-28416 Supplier Medchemexpress LLC Supplier No. HY118700A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Iminobiotin hydrobromide is a biotin analog and a reversible inhibitor of nitric oxide synthases used in research to probe NO signaling and neuroprotection. It is supplied as a 10 mM solution in DMSO (1 mL) and has reported Ki values of 21.8 μM for murine iNOS and 37.5 μM for rat n-cNOS. For research use only; not for human or therapeutic applications.

  • Biotin analog that reversibly inhibits nitric oxide synthases (NOS).
  • Reported Ki: 21.8 μM (murine iNOS) and 37.5 μM (rat n-cNOS).
  • Supplied as 10 mM solution in DMSO, 1 mL vial.
  • Molecular weight 324.24 g·mol⁻1; formula C10H18BrN3O2S.
  • Purity: 98.0%.
  • Suitable for research on NO signaling, immunology, and neuroprotection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.