Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-iminobiotin hydrobromide | 76985-52-9 | 98.0% | 324.24 g/mol | C10H18BrN3O2S | 25 MG
SDP

Catalog No. 5000226409
Encompass_Preferred

2-Iminobiotin hydrobromide (CAS 76985-52-9) is a synthetic biotin analog and a reversible inhibitor of nitric oxide synthases (iNOS and nNOS). Supplied as a solid or as ready-to-use DMSO solutions, it is used in biochemical and cellular assays to modulate nitric oxide production and investigate NOS-related pathways.

  • Reversible inhibition of iNOS and nNOS for targeted enzyme studies.
  • Suitable for biochemical assays and cellular research.
  • Available in solid form and as DMSO solutions for flexible dosing.
  • High purity (≈98.0%) to support reproducible results.
  • Molecular weight 324.24 g/mol and well-defined chemical identity (C10H18BrN3O2S).

Catalog No. 50-002-26409 Supplier Medchemexpress LLC Supplier No. HY118700A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Iminobiotin hydrobromide (CAS 76985-52-9) is a synthetic biotin analog and a reversible inhibitor of nitric oxide synthases (iNOS and nNOS). Supplied as a solid or as ready-to-use DMSO solutions, it is used in biochemical and cellular assays to modulate nitric oxide production and investigate NOS-related pathways.

  • Reversible inhibition of iNOS and nNOS for targeted enzyme studies.
  • Suitable for biochemical assays and cellular research.
  • Available in solid form and as DMSO solutions for flexible dosing.
  • High purity (≈98.0%) to support reproducible results.
  • Molecular weight 324.24 g/mol and well-defined chemical identity (C10H18BrN3O2S).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.