Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-iminobiotin hydrobromide | 76985-52-9 | MFCD00066150 | 324.24 | C10H18BrN3O2S | 50 MG
SDP

Catalog No. 5000227875
Encompass_Preferred

2-Iminobiotin hydrobromide is a biotin analogue and a reversible inhibitor of nitric oxide synthases (NOS), used in research to probe NOS-related pathways and neuroprotection mechanisms. It is supplied as the hydrobromide salt and is intended for laboratory research use only.

  • Reversible nitric oxide synthase inhibitor for iNOS and n-cNOS.
  • Used in neuroscience and neuromodulation research.
  • Supplied as a hydrobromide salt for the active molecule.
  • Molecular weight 324.24 g/mol; formula C10H18BrN3O2S.
  • Available in small research pack sizes, including 50 MG.

Catalog No. 50-002-27875 Supplier Medchemexpress LLC Supplier No. HY118700A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Iminobiotin hydrobromide is a biotin analogue and a reversible inhibitor of nitric oxide synthases (NOS), used in research to probe NOS-related pathways and neuroprotection mechanisms. It is supplied as the hydrobromide salt and is intended for laboratory research use only.

  • Reversible nitric oxide synthase inhibitor for iNOS and n-cNOS.
  • Used in neuroscience and neuromodulation research.
  • Supplied as a hydrobromide salt for the active molecule.
  • Molecular weight 324.24 g/mol; formula C10H18BrN3O2S.
  • Available in small research pack sizes, including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.