Learn More
Bioworld 2-IMINOBIOTIN SEPAROPOREA
Supplier:  Bioworld 201810561
Encompass
2-IMINOBIOTIN SEPAROPORE (AGAROSE) 6B-CL: Application: For the purification of avidin containing proteins. Specifications: Ligand: 2-Iminobiotin; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Cross-linked...
Catalog No. 50-197-807