Learn More
Medchemexpress LLC 2-Iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 137.63 | C4H8ClNS | 100 MG
SDP
Catalog No. 5000300328
Encompass_Preferred
2-Iminothiolane hydrochloride is a thiolating reagent that converts primary amines into free sulfhydryl (-SH) groups for protein, peptide, and oligosaccharide modification. It reacts under mild conditions (pH 7-10) to form an amidine linkage while introducing a reactive thiol, enabling subsequent labeling, crosslinking, or conjugation for research applications.
  • Introduces free sulfhydryl groups to primary amines.
  • Reacts efficiently at neutral to slightly basic pH (7-10).
  • Forms stable amidine linkages upon conjugation.
  • Enables protein and peptide labeling and crosslinking.
  • Suitable for bioconjugation and assay development.
Catalog No. 50-003-00328 Supplier Medchemexpress LLC Supplier No. HYW015798100MG
2-Iminothiolane hydrochloride is a thiolating reagent that converts primary amines into free sulfhydryl (-SH) groups for protein, peptide, and oligosaccharide modification. It reacts under mild conditions (pH 7-10) to form an amidine linkage while introducing a reactive thiol, enabling subsequent labeling, crosslinking, or conjugation for research applications.
  • Introduces free sulfhydryl groups to primary amines.
  • Reacts efficiently at neutral to slightly basic pH (7-10).
  • Forms stable amidine linkages upon conjugation.
  • Enables protein and peptide labeling and crosslinking.
  • Suitable for bioconjugation and assay development.