Learn More
Medchemexpress LLC 2-iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 99.9% | 137.63 | C4H8ClNS | 10 MG
SDP
Catalog No. 5000301541
Encompass_Preferred
2-Iminothiolane hydrochloride is a thiolating reagent used to introduce sulfhydryl (-SH) groups into primary amines of proteins, peptides, and oligosaccharides. It reacts under near-physiological conditions to expose free thiols for subsequent conjugation, crosslinking, or labeling in bioconjugation workflows.
  • Reacts with primary amines at pH 7-10 to generate free thiols.
  • Forms an amidine linkage that tethers the thiol to the substrate.
  • Enables protein and peptide modification for bioconjugation.
  • Suitable for glycan labeling and oligosaccharide modification.
  • High purity for reproducible experimental results (>99.9%).
  • Solid, white to off-white material that is easy to handle and weigh.
  • Available in small research-scale pack sizes for laboratory use.
Catalog No. 50-003-01541 Supplier Medchemexpress LLC Supplier No. HYW01579810MG
2-Iminothiolane hydrochloride is a thiolating reagent used to introduce sulfhydryl (-SH) groups into primary amines of proteins, peptides, and oligosaccharides. It reacts under near-physiological conditions to expose free thiols for subsequent conjugation, crosslinking, or labeling in bioconjugation workflows.
  • Reacts with primary amines at pH 7-10 to generate free thiols.
  • Forms an amidine linkage that tethers the thiol to the substrate.
  • Enables protein and peptide modification for bioconjugation.
  • Suitable for glycan labeling and oligosaccharide modification.
  • High purity for reproducible experimental results (>99.9%).
  • Solid, white to off-white material that is easy to handle and weigh.
  • Available in small research-scale pack sizes for laboratory use.