Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 99.9% | 137.63 g·mol⁻¹ | C4H8ClNS | 250 MG
SDP

Catalog No. 5000302842
Encompass_Preferred

2-Iminothiolane hydrochloride is a thiolating reagent (Traut's reagent) used to introduce sulfhydryl (-SH) groups into primary amines on proteins, peptides, and oligosaccharides for bioconjugation, labeling, and cross-linking. It reacts efficiently near neutral to slightly alkaline pH to form thiol-modified biomolecules while minimizing disruption of native structure.

  • Reacts with primary amines at pH 7-10 to generate free sulfhydryl groups.
  • Suitable for protein, peptide, and oligosaccharide modification for bioconjugation.
  • High purity (≈99.9%) for reliable, reproducible results.
  • Soluble in water (100 mg/mL) and DMSO (14.29 mg/mL) with ultrasonic or gentle warming as needed.
  • Recommended storage sealed at 4°C; prepared solutions stable at -20°C (short term) or -80°C (long term) per datasheet.

Catalog No. 50-003-02842 Supplier Medchemexpress LLC Supplier No. 250MGHYW015798
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Iminothiolane hydrochloride is a thiolating reagent (Traut's reagent) used to introduce sulfhydryl (-SH) groups into primary amines on proteins, peptides, and oligosaccharides for bioconjugation, labeling, and cross-linking. It reacts efficiently near neutral to slightly alkaline pH to form thiol-modified biomolecules while minimizing disruption of native structure.

  • Reacts with primary amines at pH 7-10 to generate free sulfhydryl groups.
  • Suitable for protein, peptide, and oligosaccharide modification for bioconjugation.
  • High purity (≈99.9%) for reliable, reproducible results.
  • Soluble in water (100 mg/mL) and DMSO (14.29 mg/mL) with ultrasonic or gentle warming as needed.
  • Recommended storage sealed at 4°C; prepared solutions stable at -20°C (short term) or -80°C (long term) per datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.