Learn More
Medchemexpress LLC 2-iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 99.9% | 137.63 g/mol | C4H8ClNS | 5 MG
SDP
Catalog No. 5000300685
Encompass_Preferred
2-Iminothiolane hydrochloride (Traut's reagent) is a thiolating reagent that converts primary amines into sulfhydryl (-SH) groups via amidine formation, enabling modification of proteins, peptides, and glycans for downstream conjugation and crosslinking. It is provided as a high-purity solid with characterized solubility and storage recommendations for biochemical applications.
  • Introduces free sulfhydryl groups into primary amines.
  • Enables protein, peptide, and glycan conjugation and crosslinking.
  • High purity suitable for biochemical assays.
  • Soluble in water and DMSO under specified conditions.
  • Stable when stored sealed at recommended temperatures.
Catalog No. 50-003-00685 Supplier Medchemexpress LLC Supplier No. HYW0157985MG
2-Iminothiolane hydrochloride (Traut's reagent) is a thiolating reagent that converts primary amines into sulfhydryl (-SH) groups via amidine formation, enabling modification of proteins, peptides, and glycans for downstream conjugation and crosslinking. It is provided as a high-purity solid with characterized solubility and storage recommendations for biochemical applications.
  • Introduces free sulfhydryl groups into primary amines.
  • Enables protein, peptide, and glycan conjugation and crosslinking.
  • High purity suitable for biochemical assays.
  • Soluble in water and DMSO under specified conditions.
  • Stable when stored sealed at recommended temperatures.