Learn More
Medchemexpress LLC 2-iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 99.9% | 137.63 g/mol | C4H8ClNS | 50 MG
SDP
Catalog No. 5000300684
Encompass_Preferred
2-Iminothiolane hydrochloride is a thiolating reagent that introduces sulfhydryl (-SH) groups into primary amines of proteins, peptides, and oligosaccharides for bioconjugation and cross-linking. Supplied as a white to off-white solid, it is intended for research use and should be stored sealed and away from moisture.
  • High purity suitable for biochemical applications
  • Efficient thiolation of primary amines under mild conditions
  • Useful for protein and peptide modification and cross-linking
  • Solid form easy to weigh and dissolve for reactions
  • Stable when stored sealed and protected from moisture
Catalog No. 50-003-00684 Supplier Medchemexpress LLC Supplier No. HYW01579850MG
2-Iminothiolane hydrochloride is a thiolating reagent that introduces sulfhydryl (-SH) groups into primary amines of proteins, peptides, and oligosaccharides for bioconjugation and cross-linking. Supplied as a white to off-white solid, it is intended for research use and should be stored sealed and away from moisture.
  • High purity suitable for biochemical applications
  • Efficient thiolation of primary amines under mild conditions
  • Useful for protein and peptide modification and cross-linking
  • Solid form easy to weigh and dissolve for reactions
  • Stable when stored sealed and protected from moisture