Learn More
Medchemexpress LLC 2-iminothiolane hydrochloride | 4781-83-3 | MFCD00039013 | 99.9% | 137.63 g·mol⁻¹ | C4H8ClNS | 500 MG
SDP
Catalog No. 5000300020
Encompass_Preferred
2-Iminothiolane hydrochloride is a thiolation reagent that introduces free sulfhydryl (-SH) groups into primary amines on proteins, peptides, and oligosaccharides under mild aqueous conditions (pH 7-10). It is commonly used to generate thiol handles for downstream conjugation, crosslinking, and labeling workflows where efficient coupling to thiol-reactive probes is required.
  • High purity suitable for biochemical applications.
  • Introduces free sulfhydryl groups to primary amines at pH 7-10.
  • Enables efficient conjugation to maleimide or thiol-reactive probes.
  • Reactive under mild, aqueous conditions preserving biomolecule integrity.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-00020 Supplier Medchemexpress LLC Supplier No. HYW015798500MG
2-Iminothiolane hydrochloride is a thiolation reagent that introduces free sulfhydryl (-SH) groups into primary amines on proteins, peptides, and oligosaccharides under mild aqueous conditions (pH 7-10). It is commonly used to generate thiol handles for downstream conjugation, crosslinking, and labeling workflows where efficient coupling to thiol-reactive probes is required.
  • High purity suitable for biochemical applications.
  • Introduces free sulfhydryl groups to primary amines at pH 7-10.
  • Enables efficient conjugation to maleimide or thiol-reactive probes.
  • Reactive under mild, aqueous conditions preserving biomolecule integrity.
  • Available in multiple pack sizes for scale flexibility.