Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iminothiolane, MP Biomedicals™

Catalog No. ICN15131980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15131980 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15131980 Supplier MP Biomedicals, Inc Supplier No. 0215131980
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4781-83-3
Molecular Formula C4H8ClNS
Molecular Weight (g/mol) 137.625
InChI Key ATGUDZODTABURZ-UHFFFAOYSA-N
Synonym 2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
PubChem CID 13166855
IUPAC Name thiolan-2-imine;hydrochloride
SMILES C1CC(=N)SC1.Cl

Specifications

Color White
Assay Percent Range 0.98
Quantity 100 mg
Formula Weight 137.3
Physical Form Crystalline Powder
Chemical Name or Material 2-Iminothiolane
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.