Learn More
Optimize Technologies 2-in-1 Seal Cap for Agilent 1050, 1100, 1200 (replaces OEM Gold Seal and Outlet Cap), 4pk
SDP
Catalog No. NC0211436
Encompass_Preferred
Our 2-In-1 Seal Cap replaces both the OEM outlet gold seal and outlet cap with a single component. Use it on your purge valve, on your OEM inlet and outlet check valves, or anywhere else on your 1200/ 1100/ 1050 system where gold seals and outlet caps are required. Please note- this item is not required with OPTI-MAX™ check valves. This item is equivalent to OEM part number 5067-4728.
Catalog No. NC0211436 Supplier Optimize Technologies Supplier No. 273103075
Our 2-In-1 Seal Cap replaces both the OEM outlet gold seal and outlet cap with a single component. Use it on your purge valve, on your OEM inlet and outlet check valves, or anywhere else on your 1200/ 1100/ 1050 system where gold seals and outlet caps are required. Please note- this item is not required with OPTI-MAX™ check valves. This item is equivalent to OEM part number 5067-4728.