Learn More
MSE Supplies LLC 2 IN SAPPHIRE WAFER R-PLANE (1
SDP
Catalog No. NC2168944
Encompass
2 in Sapphire Wafer R-Plane (1-102) Single Crystal Al2O3 single side polished
This item is not returnable. View return policy
Catalog No. NC2168944 Supplier MSE Supplies LLC Supplier No. WA0415
2 in Sapphire Wafer R-Plane (1-102) Single Crystal Al2O3 single side polished