Learn More
Grainger Hyde® Putty Knife, Brass, Stiff, 2 in. Blade, 1 ea
Catalog No. NC2603815
Encompass_Preferred
This non-sparking chisel scraper features a 2 in. stiff brass blade designed for scraping tasks and surface preparation in hazardous environments. The tool utilizes full-tang construction for enhanced strength and reliability during rigorous use.
  • Non-spark brass blade ideal for use in potentially volatile situations and on delicate surfaces
  • Stiff blade design ensures effective scraping and precise application in tight areas
  • Durable nylon handle resists most common paint thinners and solvents for long-lasting use
  • Full-tang construction backed by a lifetime guarantee against manufacturer defects
Catalog No. NC2603815 Supplier Grainger Supplier No. 5LL96
This non-sparking chisel scraper features a 2 in. stiff brass blade designed for scraping tasks and surface preparation in hazardous environments. The tool utilizes full-tang construction for enhanced strength and reliability during rigorous use.
  • Non-spark brass blade ideal for use in potentially volatile situations and on delicate surfaces
  • Stiff blade design ensures effective scraping and precise application in tight areas
  • Durable nylon handle resists most common paint thinners and solvents for long-lasting use
  • Full-tang construction backed by a lifetime guarantee against manufacturer defects