Learn More
Andwin Scientific 2 INCH STORAGE BOXES
SDP
Catalog No. NC9804321
Encompass_Preferred
Storage Box, 5cm in height. 13.3 x 13.3cm. With 81-cell divider. For use with Signature and Ultra-Low Temperature Freezers.
Catalog No. NC9804321 Supplier Andwin Scientific Supplier No. 25000134
Storage Box, 5cm in height. 13.3 x 13.3cm. With 81-cell divider. For use with Signature and Ultra-Low Temperature Freezers.