Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Indanol, 99%, Thermo Scientific™

Catalog No. AC177410250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC177410250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC177410250 Supplier Thermo Scientific Chemicals Supplier No. 177410250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4254-29-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
InChI Key KMGCKSAIIHOKCX-UHFFFAOYSA-N
Synonym 2-indanol, indan-2-ol, 2-hydroxyindan, 2-hydroxyhydrindene, 2-hydroxyindane, 1h-inden-2-ol, 2,3-dihydro, 2-indanol;, pubchem9652, acmc-1asoy
PubChem CID 77936
IUPAC Name 2,3-dihydro-1H-inden-2-ol
SMILES C1C(CC2=CC=CC=C21)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 65°C to 70°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥98.5%
Physical Form Needle-like Powder
Chemical Name or Material 2-Indanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.