Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Iodo-1-methyl-1H-imidazole

Catalog No. CDS005078 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005078 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005078 Supplier Sigma Aldrich Supplier No. CDS005078100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • α-nonanoic lactone has been used:
  • In quantitative analysis of oak volatile compounds in aged red wine by headspace solid-phase microextraction-GC-ms method
  • As substrate in serum paraoxonase-1 (pon1) enzyme assay
  • In synthesis of 4-hydroxynonanal via reduction of α-nonanoic lactone by diisobutylaluminum hydride in toluene

Specifications

CAS 37067-95-1
Molecular Formula C4H5IN2
Linear Formula C4H5IN2
MDL Number MFCD00661606
Quantity 100 mL
Molecular Weight (g/mol) 208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.