Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Iodo-5-methoxypyridine

Catalog No. SYX003201G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003201G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003201G Supplier Sigma Aldrich Supplier No. SYX003201G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,2,2-trichloroethyl chloroformate was used as:
  • Derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples
  • Starting reagent during the synthesis of 6-nor-9,10-dihydrolysergic acid methyl ester (iv)
  • Reagent during n-demethylation of dextromethorphan
  • Protecting group reagent for amines and alcohols

Specifications

CAS 163129-79-1
Molecular Formula C6H6INO
Linear Formula C6H6INO
MDL Number MFCD02259696
Quantity 100 mL
Molecular Weight (g/mol) 235.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.