Learn More
Medchemexpress LLC 2-Iodoacetamide | 144-48-9 | MFCD00008028 | ≥99% | 184.96 | 1 G
SDP
Catalog No. 5000404636
Encompass_Preferred
2-Iodoacetamide is an alkylating agent commonly used for the alkylation of cysteine during sample preparation for proteomics. It is also known to produce chronic deep ulcers in the secretory part of the rat stomach when administered.
  • Commonly used for alkylation of cysteine
  • Used during sample preparation for proteomics
  • Functions as a DNA alkylator/crosslinker
Catalog No. 50-004-04636 Supplier Medchemexpress LLC Supplier No. HY344771G
2-Iodoacetamide is an alkylating agent commonly used for the alkylation of cysteine during sample preparation for proteomics. It is also known to produce chronic deep ulcers in the secretory part of the rat stomach when administered.
  • Commonly used for alkylation of cysteine
  • Used during sample preparation for proteomics
  • Functions as a DNA alkylator/crosslinker