Learn More
Medchemexpress LLC HY-34477 500mg Medchemexpress, 2-Iodoacetamide CAS:144-48-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY34477500MG
Encompass_Preferred
Medchemexpress, HY-34477 500mg 2-Iodoacetamide CAS:144-48-9 2-Iodoacetamide (Iodoacetamide), an alkylating agent, is a commonly used agent for alkylation of cysteine during sample preparation for proteomics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2234