Learn More
Medchemexpress LLC 2-iodoacetamide-d4 | 1219802-64-8 | MFCD06658608 | 99.9% | 188.99 | C2D4INO | 10 MG
SDP
Catalog No. 5000296776
Encompass_Preferred
2-Iodoacetamide-d4 is the deuterium-labeled analog of 2-iodoacetamide, an alkylating reagent commonly used to modify cysteine residues during proteomics sample preparation. The labeled compound is used as an internal standard or tracer in mass spectrometry workflows and has specified stability and storage recommendations for powdered and solution forms.
  • Deuterium-labeled analog for isotope-dilution MS.
  • Suitable for cysteine alkylation in proteomics sample preparation.
  • High purity (99.9%) appropriate for analytical use.
  • Small pack sizes available for method development.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-96776 Supplier Medchemexpress LLC Supplier No. HY34477S110MG
2-Iodoacetamide-d4 is the deuterium-labeled analog of 2-iodoacetamide, an alkylating reagent commonly used to modify cysteine residues during proteomics sample preparation. The labeled compound is used as an internal standard or tracer in mass spectrometry workflows and has specified stability and storage recommendations for powdered and solution forms.
  • Deuterium-labeled analog for isotope-dilution MS.
  • Suitable for cysteine alkylation in proteomics sample preparation.
  • High purity (99.9%) appropriate for analytical use.
  • Small pack sizes available for method development.
  • Stable when stored under recommended conditions.