Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Iodoaniline | 615-43-0 | MFCD00007680 | 219.03 | 1 G
SDP

Catalog No. 5000460268
Encompass_Preferred

2-Iodoaniline (2-Iodophenylamine) is an aniline derivative known for its potential hepatotoxic and nephrotoxic activity. Studies show it can induce oliguria, diminish kidney weight, cause tubular casts, and alter morphology in the centrilobular region in male Fischer 344 rats. This product is intended for research use only.

  • Molecular formula: C6H6IN
  • CAS number: 615-43-0
  • Molecular weight: 219.03
  • Appearance: Solid
  • Color: White to light brown
  • Solubility in DMSO: 100 mg/mL (Need ultrasonic)
  • Solid storage: 4°C, protected from light
  • In solvent storage: -80°C for 6 months, -20°C for 1 month, protected from light
  • Purity: 99.97%
  • Target: Drug derivative

Catalog No. 50-004-60268 Supplier Medchemexpress LLC Supplier No. HYW0073181G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Iodoaniline (2-Iodophenylamine) is an aniline derivative known for its potential hepatotoxic and nephrotoxic activity. Studies show it can induce oliguria, diminish kidney weight, cause tubular casts, and alter morphology in the centrilobular region in male Fischer 344 rats. This product is intended for research use only.

  • Molecular formula: C6H6IN
  • CAS number: 615-43-0
  • Molecular weight: 219.03
  • Appearance: Solid
  • Color: White to light brown
  • Solubility in DMSO: 100 mg/mL (Need ultrasonic)
  • Solid storage: 4°C, protected from light
  • In solvent storage: -80°C for 6 months, -20°C for 1 month, protected from light
  • Purity: 99.97%
  • Target: Drug derivative

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.