Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. I06355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06355G 5 g
I06351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06355G Supplier TCI America Supplier No. I06355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40400-13-3
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00236046
InChI Key GQFITODJWOIYPF-UHFFFAOYSA-N
Synonym 2-iodobenzyl bromide, 1-bromomethyl-2-iodobenzene, o-iodobenzyl bromide, alpha-bromo-2-iodotoluene, benzene, 1-bromomethyl-2-iodo, 2-iodobenzylbromide, 2-iodo benzylbromide, pubchem14812, acmc-1anor, alpha-bromo-2-iodo toluene
PubChem CID 11426472
IUPAC Name 1-(bromomethyl)-2-iodobenzene
SMILES C1=CC=C(C(=C1)CBr)I

Specifications

Melting Point 55°C
Boiling Point 125°C
Quantity 5 g
UN Number 3261
Formula Weight 296.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.