Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodocumene 98.0+%, TCI America™
SDP

Catalog No. I05595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05595G Supplier TCI America Supplier No. I05595G
Add to Cart
Add to Cart

Specifications

CAS 19099-54-8
Boiling Point 117°C
Molecular Formula C9H11I
Quantity 5 g
MDL Number MFCD00013704
Synonym 1-iodo-2-isopropylbenzene, 2-iodoisopropylbenzene, 2-iodocumene, o-iodocumene, 1-iodo-2-isopropyl-benzene, benzene, 1-iodo-2-1-methylethyl, o-i-propyliodobenzene, 2-isopropyliodobenzene, 2-iodoisopropyl benzene, pubchem12584
InChI Key SORQIYFSJAWBNQ-UHFFFAOYSA-N
SMILES CC(C)C1=CC=CC=C1I
IUPAC Name 1-iodo-2-(propan-2-yl)benzene
Molecular Weight (g/mol) 246.09
PubChem CID 87931
Formula Weight 246.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodocumene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.