Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Iodoethanol

Catalog No. 1768505G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-685-05G 5 g
17-685-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-685-05G Supplier Sigma Aldrich Supplier No. 1768505G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The standard molar enthalpy of 2-iodoethanol formation has been studied.2-Iodoethanol was used in the synthesis of the neuroexcitants 3-(carboxymethyl)pyrrolidine-2,4-dicarboxylic acid, α-kainic acid, alpha-isokainic acid and alpha-dihydroallokainic acid.

Specifications

CAS 624-76-0
Density 2.205 g/mL (at 25°C (literature))
Boiling Point 85°C (25 mmHg)
Molecular Formula C2H5IO
Refractive Index n20/D 1.572 (literature)
Linear Formula ICH2CH2OH
MDL Number MFCD00002830
Quantity 5 g
Molecular Weight (g/mol) 171.97
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.