Learn More
Eurofins Discoverx 2-Iodomelatonin, 10 mg. (92-1047)
Supplier:  Eurofins Discoverx 921047
Encompass
2-iodomelatonin is a melatonin analog and potent agonist of the melatonin receptors MT1 and MT2 (pKi 10.55 and 9.87, respectively). It Inhibits receptor-mediated cAMP accumulation and has been used to characterize melatonin binding sites in the brain and peripheral tissues. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9590