Learn More
Medchemexpress LLC HY-101176 5mg Medchemexpress, 2-Iodomelatonin CAS:93515-00-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011765MG
Encompass_Preferred
Medchemexpress, HY-101176 5mg 2-Iodomelatonin CAS:93515-00-5 2-Iodomelatonin is a potent agonist of melatonin receptor 1 (MT1) with a Ki value of 28 pM, it is more 5-fold selective for MT1  over MT2. 2-iodomelatonin can be used to identify, characterize and localize melatonin binding sites in the brain and peripheral tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7809