Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Iodoxybenzoic Acid

Catalog No. 6613841G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-138-41G 1 g
66-138-410G 10 g
66-138-4250G 250 g
66-138-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 66-138-41G Supplier Sigma Aldrich Supplier No. 6613841G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Stabilized 2-Iodoxybenzoic Acid (SIBX)The material is stabilized with benzoic acid and isophthalic acid

Specifications

CAS 61717-82-6
Molecular Formula C7H5IO4
Linear Formula C7H5IO4
MDL Number MFCD02912492
Quantity 1 g
Synonym SIBX; Stabilized IBX
Molecular Weight (g/mol) 280.02 g/mol
Biological Activity Lungs, Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.