Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodoxybenzoic Acid (stabilized with Benzoic Acid + Isophthalic Acid) 39.0+%, TCI America™
SDP

Catalog No. I079125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I079125G 25 g
I07915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I079125G Supplier TCI America Supplier No. I079125G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61717-82-6
Molecular Formula C7H5IO4
MDL Number MFCD02912492
Synonym IBX

Specifications

Melting Point 232°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 280.02
Percent Purity ≥39.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodoxybenzoic Acid (stabilized with Benzoic Acid + Isophthalic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.