Learn More
Medchemexpress LLC 2-isobutyl-3-methoxypyrazine | 24683-00-9 | MFCD00006128 | 99.8% | 166.22 g/mol | C9H14N2O | 1 ML
SDP
Catalog No. 5000299766
Encompass_Preferred
2-Isobutyl-3-methoxypyrazine is a volatile pyrazine present in grapes, wines, green pepper, and asparagus, supplied as a research-grade analytical standard and solution for aroma and flavor studies. It is provided with supporting documentation for laboratory use and storage guidance.
  • High purity (99.83%) suitable for analytical applications.
  • Available as a concentrated DMSO solution for convenient dosing.
  • Soluble in DMSO at high concentration (≈300.8 mM) for stock preparation.
  • Colorless to light yellow liquid with defined density.
  • Long shelf life when stored per recommended conditions.
  • Provided with COA, SDS, and datasheet documentation.
Catalog No. 50-002-99766 Supplier Medchemexpress LLC Supplier No. HYW01714110MM/1ML
2-Isobutyl-3-methoxypyrazine is a volatile pyrazine present in grapes, wines, green pepper, and asparagus, supplied as a research-grade analytical standard and solution for aroma and flavor studies. It is provided with supporting documentation for laboratory use and storage guidance.
  • High purity (99.83%) suitable for analytical applications.
  • Available as a concentrated DMSO solution for convenient dosing.
  • Soluble in DMSO at high concentration (≈300.8 mM) for stock preparation.
  • Colorless to light yellow liquid with defined density.
  • Long shelf life when stored per recommended conditions.
  • Provided with COA, SDS, and datasheet documentation.