Learn More
Medchemexpress LLC 2-isobutyl-3-methoxypyrazine-d3 | 588732-63-2 | >98.0% | 169.24 g/mol | C9H11D3N2O | 10 MG
SDP
Catalog No. 5000299515
Encompass_Preferred
2-Isobutyl-3-methoxypyrazine-d3 is the deuterium-labeled analogue of 2-isobutyl-3-methoxypyrazine, an aroma constituent in grapes, wines, green pepper, and asparagus. The labeled material (d3) is provided as a reference standard for analytical and research use, including isotope-dilution assays, method development, and trace-level quantitation.
Catalog No. 50-002-99515 Supplier Medchemexpress LLC Supplier No. HYW017141S10MG
2-Isobutyl-3-methoxypyrazine-d3 is the deuterium-labeled analogue of 2-isobutyl-3-methoxypyrazine, an aroma constituent in grapes, wines, green pepper, and asparagus. The labeled material (d3) is provided as a reference standard for analytical and research use, including isotope-dilution assays, method development, and trace-level quantitation.