Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isobutylthiazole, 99%

Catalog No. AAA1375914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1375914 25 g
AAA1375906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1375914 Supplier Thermo Scientific Chemicals Supplier No. A1375914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18640-74-9 | C7H11NS | 141.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18640-74-9
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00005334
InChI Key CMPVUVUNJQERIT-UHFFFAOYSA-N
Synonym 2-isobutylthiazole, 2-isobutyl-1,3-thiazole, 2-2-methylpropyl thiazole, thiazole, 2-isobutyl, 2-2-methylpropyl-1,3-thiazole, 2-isobutyl thiazole, thiazole, 2-2-methylpropyl, iso-butyl thiazole, fema no. 3134
PubChem CID 62725
IUPAC Name 2-(2-methylpropyl)-1,3-thiazole
SMILES CC(C)CC1=NC=CS1

Specifications

Density 0.998
Boiling Point 172°C to 173°C
Flash Point 58°C (136°F)
Odor Characteristic
Refractive Index 1.496
Quantity 25 g
UN Number UN1993
Beilstein 507823
Formula Weight 141.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Isobutylthiazole
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement

H226-H315-H319-H335

Flammable liquid and vapour.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-470-1
RTECSNumber XJ5103412
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.