Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Isobutylthio)ethanol 98.0+%, TCI America™
SDP

Catalog No. I02645ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02645ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02645ML Supplier TCI America Supplier No. I02645ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42779-10-2
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.237
MDL Number MFCD00014040
InChI Key PSCYOFJZGIAXOL-UHFFFAOYSA-N
Synonym 2-Hydroxyethyl Isobutyl Sulfide
PubChem CID 545870
IUPAC Name 2-(2-methylpropylsulfanyl)ethanol
SMILES CC(C)CSCCO

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 mL
Formula Weight 134.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Isobutylthio)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.