Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropenyl-5-methyl-5-vinyltetrahydrofuran (mixture of isomers) 97.0+%, TCI America™
SDP

Catalog No. I074425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I074425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I074425G Supplier TCI America Supplier No. I074425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13679-86-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00065422
InChI Key XIGFNCYVSHOLIF-UHFFFAOYSA-N
Synonym Dehydroxylinalool Oxide
PubChem CID 61665
IUPAC Name 2-ethenyl-2-methyl-5-prop-1-en-2-yloxolane
SMILES CC(=C)C1CCC(O1)(C)C=C

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 55°C
Formula Weight 152.24
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2-Isopropenyl-5-methyl-5-vinyltetrahydrofuran (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.