Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Isopropoxyethanol

Catalog No. 1078912L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-789-12L 2 L
10-789-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-789-12L Supplier Sigma Aldrich Supplier No. 1078912L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-isopropoxyethanol molecules self-associate by hydrogen bonding between oh group and partially etheric o atom and also by hydrophobic interaction between isopropoxyethyl groups.2-isopropoxyethanol was used to study the products of the gas-phase reactions of oh radical with n-butyl methyl ether and 2-isopropoxyethanol in the presence of no
  • 2-isopropoxyethanol was used to develop methodology for evaluating electromagnetic sensor performance in nonrelaxing and nonconducting (e.g., sandy soil) media

Specifications

CAS 109-59-1
Density 0.903 g/mL at 25°C (lit.)
Boiling Point 42°C to 44°°C (13 mmHg)
Molecular Formula C5H12O2
Refractive Index n20/D 1.41 (lit.)
Linear Formula (CH3)2CHOCH2CH2OH
MDL Number MFCD00002866
Quantity 2 L
Synonym Ethylene glycol monoisopropyl ether; Isopropylglycol
Molecular Weight (g/mol) 104.15
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.