Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropyl-2-methacryloyloxyadamantane 98.0+%, TCI America™
SDP

Catalog No. I09625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09625G 5 g
I096225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09625G Supplier TCI America Supplier No. I09625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 297156-50-4
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.393
MDL Number MFCD22571643
InChI Key ZMAOPHHNBQIJOQ-UHFFFAOYSA-N
Synonym 2-Isopropyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Isopropyl-2-adamantyl Ester
PubChem CID 12097425
IUPAC Name (2-propan-2-yl-2-adamantyl) 2-methylprop-2-enoate
SMILES CC(C)C1(C2CC3CC(C2)CC1C3)OC(=O)C(=C)C

Specifications

Melting Point 40°C
Color White
Quantity 5 g
Formula Weight 262.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropyl-2-methacryloyloxyadamantane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.