Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylaniline 98.0+%, TCI America™
SDP

Catalog No. I031825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I031825ML 25 mL
I0318250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I031825ML Supplier TCI America Supplier No. I031825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 643-28-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007720
InChI Key YKOLZVXSPGIIBJ-UHFFFAOYSA-N
Synonym 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline
PubChem CID 12561
IUPAC Name 2-(propan-2-yl)aniline
SMILES CC(C)C1=CC=CC=C1N

Specifications

Boiling Point 104°C
Quantity 25 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Isopropylaniline
EINECSNumber (3)-0206
RTECSNumber BY4200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.