Learn More
Medchemexpress LLC 2-Isopropylbenzeneboronic acid | 89787-12-2 | 164.01 | 10 G
SDP
Catalog No. 5000448640
Encompass_Preferred
2-Isopropylbenzeneboronic acid (2-Isopropylphenylboronic acid) is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • High purity: typically 99.80% or 99.94%.
  • Appears as a solid, ranging from white to off-white.
  • Can be used in various life science research applications.
  • Provided with detailed documentation, including data sheet, SDS, and certificate of analysis.
  • Offers specific solubility in DMSO for in vitro applications.
  • Protocols available for in vivo dissolution using co-solvents like DMSO, PEG300, Tween-80, and saline.
Catalog No. 50-004-48640 Supplier Medchemexpress LLC Supplier No. HYW00305010G
Add to Cart
Add to Cart
2-Isopropylbenzeneboronic acid (2-Isopropylphenylboronic acid) is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.
  • High purity: typically 99.80% or 99.94%.
  • Appears as a solid, ranging from white to off-white.
  • Can be used in various life science research applications.
  • Provided with detailed documentation, including data sheet, SDS, and certificate of analysis.
  • Offers specific solubility in DMSO for in vitro applications.
  • Protocols available for in vivo dissolution using co-solvents like DMSO, PEG300, Tween-80, and saline.