Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylbenzenethiol 95.0+%, TCI America™
SDP

Catalog No. I08575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08575G Supplier TCI America Supplier No. I08575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6262-87-9
Molecular Formula C9H12S
Molecular Weight (g/mol) 152.255
MDL Number MFCD00004837
InChI Key QEDRUXIMTJVXFL-UHFFFAOYSA-N
Synonym 2-Isopropylthiophenol
PubChem CID 96003
IUPAC Name 2-propan-2-ylbenzenethiol
SMILES CC(C)C1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 227°C
Quantity 5 g
UN Number 1760
Formula Weight 152.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropylbenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.