Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylphenol, 98+%

Catalog No. p-7081856
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1202718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1202718 Supplier Thermo Scientific Chemicals Supplier No. A1202718
Only null left
Add to Cart

CAS: 88-69-7 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-69-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002224
InChI Key CRBJBYGJVIBWIY-UHFFFAOYSA-N
Synonym 2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
PubChem CID 6943
ChEBI CHEBI:38506
IUPAC Name 2-propan-2-ylphenol
SMILES CC(C)C1=CC=CC=C1O

Specifications

Melting Point 14°C to 16°C
Density 1.01
Boiling Point 212°C to 214°C
Flash Point 88°C (190°F)
Assay Percent Range 98%
Refractive Index 1.527
Quantity 50 g
UN Number UN2922
Beilstein 1363322
Sensitivity Air sensitive
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2-Isopropylphenol
Show More Show Less
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 201-852-8
RTECSNumber SL5900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.