Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-isopropylphenyl (2-(naphthalen-2-yl)ethyl)carbamate | 1354359-53-7 | MFCD20926355 | 99.5% | 333.42 | C22H23NO2 | 5 MG
SDP

Catalog No. 5000240745
Encompass_Preferred

JW480 is a potent, selective small-molecule inhibitor of the serine hydrolase KIAA1363, supplied as a high-purity research reagent for biochemical and cellular studies. It is provided as a solid or as a 10 mM solution in DMSO for in vitro applications.

  • Potent and selective inhibitor of KIAA1363.
  • High purity (about 99.5%).
  • Available as solid in multiple sizes and as a 10 mM DMSO solution.
  • Molecular formula C22H23NO2; molecular weight 333.42 g/mol.
  • Soluble in DMSO and suitable for biochemical and cellular assays.
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-40745 Supplier Medchemexpress LLC Supplier No. HY1075825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JW480 is a potent, selective small-molecule inhibitor of the serine hydrolase KIAA1363, supplied as a high-purity research reagent for biochemical and cellular studies. It is provided as a solid or as a 10 mM solution in DMSO for in vitro applications.

  • Potent and selective inhibitor of KIAA1363.
  • High purity (about 99.5%).
  • Available as solid in multiple sizes and as a 10 mM DMSO solution.
  • Molecular formula C22H23NO2; molecular weight 333.42 g/mol.
  • Soluble in DMSO and suitable for biochemical and cellular assays.
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.