Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-isopropylphenyl (2-(naphthalen-2-yl)ethyl)carbamate | 1354359-53-7 | MFCD20926355 | 99.5% | 333.42 g/mol | C22H23NO2 | 50 MG
SDP

Catalog No. 5000240744
Encompass_Preferred

JW480 is a potent, selective small-molecule inhibitor of the serine hydrolase KIAA1363 (AADACL1) used in biochemical and cell-based research to probe enzyme function and related pathways. This compound is supplied for research use only.

  • Potent, selective inhibitor of KIAA1363.
  • Intended for research use only.
  • High purity: 99.54%.
  • Chemical formula: C22H23NO2.
  • Molecular weight: 333.42 g/mol.
  • CAS number: 1354359-53-7.
  • Supplied as a 50 mg quantity.

Catalog No. 50-002-40744 Supplier Medchemexpress LLC Supplier No. HY10758250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JW480 is a potent, selective small-molecule inhibitor of the serine hydrolase KIAA1363 (AADACL1) used in biochemical and cell-based research to probe enzyme function and related pathways. This compound is supplied for research use only.

  • Potent, selective inhibitor of KIAA1363.
  • Intended for research use only.
  • High purity: 99.54%.
  • Chemical formula: C22H23NO2.
  • Molecular weight: 333.42 g/mol.
  • CAS number: 1354359-53-7.
  • Supplied as a 50 mg quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.