Learn More
Medchemexpress LLC 2-isopropylphenyl N-(2-naphthalen-2-yl)ethyl carbamate | 1354359-53-7 | MFCD20926355 | 99.5% | 333.42 g/mol | C22H23NO2 | 10 MG
SDP
Catalog No. 5000240767
Encompass_Preferred
JW480 is a potent, selective carbamate inhibitor of the serine hydrolase KIAA1363 (AADACL1). It is used as a research reagent to probe KIAA1363 function in lipid metabolism and cancer-relevant cellular pathways, and is supplied with characterization data appropriate for biochemical and cellular studies.
  • Selective inhibitor of the serine hydrolase KIAA1363.
  • Suitable for biochemical and cell-based assays.
  • Carbamate-based small-molecule structure for target engagement studies.
  • High purity and defined molecular properties for reproducible results.
  • Provided with catalog and registry identifiers for traceability.
Catalog No. 50-002-40767 Supplier Medchemexpress LLC Supplier No. HY10758210MG
JW480 is a potent, selective carbamate inhibitor of the serine hydrolase KIAA1363 (AADACL1). It is used as a research reagent to probe KIAA1363 function in lipid metabolism and cancer-relevant cellular pathways, and is supplied with characterization data appropriate for biochemical and cellular studies.
  • Selective inhibitor of the serine hydrolase KIAA1363.
  • Suitable for biochemical and cell-based assays.
  • Carbamate-based small-molecule structure for target engagement studies.
  • High purity and defined molecular properties for reproducible results.
  • Provided with catalog and registry identifiers for traceability.