Learn More
Medchemexpress LLC 2-(Isopropylthio)aniline hydrochloride | 861343-73-9 | 203.73 | 25 G

Supplier: Medchemexpress LLC 25GHYI0912
2-(Isopropylthio)aniline hydrochloride is a chemical compound identified by MedChemExpress, intended for laboratory use and substance manufacturing. This product is stable under recommended storage conditions and should be kept in a cool, well-ventilated area, away from direct sunlight and ignition sources. It is supplied in solid form.
- Identified as a laboratory chemical
- Stable under recommended storage conditions
- Should be stored in a cool, well-ventilated area
- Shipping at room temperature is suitable for less than 2 weeks
- Available as a solid appearance
- Molecular formula: C9H14CINS
- Molecular weight: 203.73
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.