Learn More
Medchemexpress LLC 2-Isopropylthioxanthone | 5495-84-1 | 99.95% | 254.35 | 250 MG
SDP
Catalog No. 5000366416
Encompass_Preferred
2-Isopropylthioxanthone is a photoinitiator commonly used in printing inks for packaging materials. It initiates the radical polymerization of the acrylic components in inks, which facilitates the drying process of the liquid ink film.
  • Widely used in printing ink applications.
  • Triggers radical polymerization of acrylic components.
  • Aids in drying liquid ink films.
Catalog No. 50-003-66416 Supplier Medchemexpress LLC Supplier No. 250MGHYW011875
2-Isopropylthioxanthone is a photoinitiator commonly used in printing inks for packaging materials. It initiates the radical polymerization of the acrylic components in inks, which facilitates the drying process of the liquid ink film.
  • Widely used in printing ink applications.
  • Triggers radical polymerization of acrylic components.
  • Aids in drying liquid ink films.