Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Keto-3-deoxyoctonate ammonium salt

Catalog No. K27555MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K27555MG 5 mg
K275550MG 50 mg
K275510MG 10 mg
K27551MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K27555MG Supplier Sigma Aldrich Supplier No. K27555MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

KDO is a sialic acid that is a component of bacterial lipopolysaccharides and a possible target for antibiotic development.

Specifications

CAS 103404-70-2
Molecular Formula C8H14O8 -+ H3N
Linear Formula C8H14O8 -+ NH3
MDL Number MFCD00057549
Quantity 5 mg
Synonym 3-Deoxyoctulosonic acid; KDO
Molecular Weight (g/mol) 255.22
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.