Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Keto-D-gluconic acid, hemicalcium salt monohydrate, >98%, MP Biomedicals™

Catalog No. ICN10036501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10036501 1 g
ICN10036505 5 g
ICN10036510 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10036501 Supplier MP Biomedicals, Inc Supplier No. 0210036501
Add to Cart
Add to Cart

2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3470-37-9
Molecular Formula C12H20CaO14
Molecular Weight (g/mol) 428.356
MDL Number MFCD00149104
InChI Key QACANIDDQFGAJU-JQTJLMCZSA-N
Synonym calcium 2-keto-d-gluconate
PubChem CID 131852723
IUPAC Name calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
SMILES C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]

Specifications

Color White
Assay Percent Range >98%
Quantity 1 g
Formula Weight 213.2
Physical Form Powder
Chemical Name or Material 2-Keto-D-gluconic acid, hemicalcium salt
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.