Learn More
Medchemexpress LLC 2-keto-D-glucose | 1854-25-7 | MFCD01863408 | 98.4% | 178.14 | C6H10O6 | 5 MG
SDP
Catalog No. 5000263625
Encompass_Preferred
2-keto-D-glucose (D-glucosone) is a small-molecule carbohydrate intermediate used as a synthetic building block and reference in biochemical research. Supplied as a solid with documented purity, molecular weight, and supporting documentation, it is suitable for analytical and synthetic applications.
  • High purity (98.4%) for reliable experimental results.
  • Solid form stable for storage and handling.
  • Well-characterized molecular properties (MW 178.14, formula C6H10O6).
  • Available documentation including COA and SDS for quality assurance.
  • Suitable as a synthetic intermediate and reference standard.
Catalog No. 50-002-63625 Supplier Medchemexpress LLC Supplier No. HY1136295MG
2-keto-D-glucose (D-glucosone) is a small-molecule carbohydrate intermediate used as a synthetic building block and reference in biochemical research. Supplied as a solid with documented purity, molecular weight, and supporting documentation, it is suitable for analytical and synthetic applications.
  • High purity (98.4%) for reliable experimental results.
  • Solid form stable for storage and handling.
  • Well-characterized molecular properties (MW 178.14, formula C6H10O6).
  • Available documentation including COA and SDS for quality assurance.
  • Suitable as a synthetic intermediate and reference standard.