Learn More
Cayman Chemical 2-keto D-Glucose, Purity: ≥95%, CAS: 1854-25-7, 50 mg
SDP
Catalog No. 502983364
Encompass_Preferred
A building block in the synthesis of carbohydrates; an intermediate in the conversion of D-glucose into D-fructose; has slow-onset, long-lasting, antioxidant properties
Catalog No. 50-298-3364 Supplier Cayman Chemical Supplier No. 2090850 MG
A building block in the synthesis of carbohydrates; an intermediate in the conversion of D-glucose into D-fructose; has slow-onset, long-lasting, antioxidant properties